missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Evaporating Dishes

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Evaporating Dishes feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 888/8

Code : 02

Additional Details : Weight : 0.58300kg

 View more versions of this product

Product Code. 12005053

  • £62.23 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Flat
200mm
55mm
Evaporating Dish
1100mL
200mm
Porcelain
Videos