Learn More
Haldenwanger™ Porcelain Evaporating Basins
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Basins feature a smooth internal finish, constructed from optimized laboratory porcelain.
Brand: Haldenwanger™ 131/6
Code : 02
Additional Details : Weight : 0.00940kg
Description
Sizes up to 175mL glazed except rim; 1,000mL and larger are glazed except outside base.Specifications
250mL | |
110mm | |
Porcelain |
110mm | |
50mm | |
Evaporating Basin |