Learn More
Haldenwanger™ Lids for Porcelain Crucibles
Use these lids with porcelain crucibles. Haldenwanger™ Lids for Porcelain Crucibles are constructed from optimized laboratory porcelain.
Brand: Haldenwanger™ 79D/8
Code : 02
Additional Details : Weight : 0.06300kg
Description
- Available in a variety of sizes
- For use with Haldenwanger porcelain crucibles
- Complies with DIN VDE 0335, type C 100
Specifications
Porcelain | |
32mm | |
Crucible Lid |