missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Casseroles

Brand:  Haldenwanger™ 1740

Code : 02

Additional Details : Weight : 0.07000kg

Product Code. 10495001

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

100mL
63mm
Porcelain
63mm
35mm
Casserole
Videos