missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Buchner Funnel

Use these funnels for a variety of filtration applications. Haldenwanger™ Porcelain Buchner Funnels are constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 127C/2

UNSPSC : 41120000

Code : 02

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12025113

  • £64.95 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

74mm
1Each
150mm
Buchner Funnel
74mm
135mL
Porcelain