missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Buchner Funnel

Use these funnels for a variety of filtration applications. Haldenwanger™ Porcelain Buchner Funnels are constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 127C/3

UNSPSC : 41120000

Code : 02

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12045113

  • £108.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

114mm
1Each
201mm
Buchner Funnel
114mm
580mL
Porcelain