missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Basins

Brand:  Haldenwanger™ 33C/4B

Code : 02

Additional Details : Weight : 0.03200kg

Product Code. 12015073

  • £11.13 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Shallow form porcelain basin
  • Available in a variety of sizes
  • Complies with DIN VDE 0335, type C 100
Specifications

Specifications

13mm
70mm
Basin
Dimension: 70 x 46 x 13mm
Porcelain
Videos