missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Basins

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Basins feature a smooth internal finish, constructed from optimized laboratory porcelain.

Brand:  Haldenwanger™ 33C/00

Code : 02

Additional Details : Weight : 0.01100kg

 View more versions of this product

Product Code. 12095063

  • £7.86 / Each


Estimated Shipment
12-02-2018
Add to basket

Description

Description

  • Shallow form porcelain basin
  • Available in a variety of sizes
  • Complies with DIN VDE 0335, type C 100
Specifications

Specifications

15mm
50mm
Basin
Dimension: 50 x 40 x 15mm
Porcelain
Videos