missing translation for 'onlineSavingsMsg'
Learn More

Ponceau S, 85%, MP Biomedicals Inc

Catalog Number p-4483053
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
6226-79-5
C22H12N4Na4O13S4
760.552
VSXKEUCERCWGKF-STNZDNLRSA-J
3-Hydroxy-4-(2-sulfo-4-(4-sulfophenylazo)phenylazo)-2, 7-naphthalene-disulfonic acid tetrasodium salt, C.I. 27195
11320219
tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
This item is not returnable. View return policy
6226-79-5
C22H12N4Na4O13S4
760.552
VSXKEUCERCWGKF-STNZDNLRSA-J
3-Hydroxy-4-(2-sulfo-4-(4-sulfophenylazo)phenylazo)-2, 7-naphthalene-disulfonic acid tetrasodium salt, C.I. 27195
11320219
tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
Assay Percent Range ∽ 85%
Formula Weight 760.6
Quantity 10 g
Chemical Name or Material Ponceau S, Protein stain for electrophoresis
compliance-icons
Product Identifier
  • Ponceau S
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'