Learn More
POMK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553037
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TSNFSILAND LDALPLVNHS SGMLVKCGHR ELHGDFVAPE QLWPYGEDVP FHDDLMPSYD EKIDIWKIPD ISSFLLGHIE GSDMVRFHLF DIHKACKSQT PSERPTAQDV LETYQKVLDT L Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
POMK Polyclonal antibody specifically detects POMK in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
POMK | |
Polyclonal | |
Unconjugated | |
POMK | |
4930444A02Rik; MDDGA12; MDDGC12; Pomk; probable inactive protein kinase-like protein SgK196; protein kinase-like protein SgK196; protein kinase-like protein SgK196-like; protein O-mannose kinase; protein-O-mannose kinase; RGD1310810; SGK196; sugen kinase 196 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84197 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H5K3 | |
POMK | |
Recombinant protein corresponding to Human POMK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only