missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000, Thermo Scientific Chemicals
130.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Beige-Brown to White |
Physical Form | Crystalline Powder |
Linear Formula | (-CH2CH(OSO3K)-)n |
Quantity | 5 g |
IUPAC Name | potassium ethenyl sulfate |
Percent Purity | ≥21.2% (K) |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Chemical Name or Material | Polyvinylsulfuric acid potassium salt |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'