missing translation for 'onlineSavingsMsg'
Learn More

Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight, Thermo Scientific Chemicals

Catalog Number p-7051455
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
9002-89-5
(C2H4O)n
44.05
MFCD00081922
IMROMDMJAWUWLK-UHFFFAOYSA-N
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
11199
ethenol
OC(-*)C-*
This item is not returnable. View return policy
9002-89-5
(C2H4O)n
44.05
MFCD00081922
IMROMDMJAWUWLK-UHFFFAOYSA-N
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
11199
ethenol
OC(-*)C-*

Polyvinyl alcohol is used in food industries as a binding and coating agent. Its viscosity favors the application as coating agent to tablets and capsules. Mixed-matrix membrane prepared by dispersion of NaA zeolite particles is used to study the pervaporative separation of water-butanol mixtures. Polyvinyl alcohol /gelatin based biocompatible polymeric scaffolds have been developed for 3D cancer models.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Solid
Solubility Information Soluble in water.
Quantity 25 g
Percent Purity 98 to 99%
Odor Odorless
Chemical Name or Material Polyvinyl alcohol, High molecular weight, Hydrolyzed

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'