missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Polytril™ Air 3/4 Gloves

Catalog Number p-7115916
Click to view available options
Size:
7
8
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Resistant polyamide knitted gloves with nitrile foam-coated palm and fingers provide good grip for precision work, especially in greasy, damp, or dirty environments. Honeywell Safety Products™ Polytril™ Air 3/4 Gloves are breathable, comfortable and provide excellent resistance.

ul>
  • Black, non-pilling polyamide knitted glove suitable for precision work.
  • Suitable for handling fine products or dirty objects in greasy, damp or dirty environments.
  • Excellent abrasion and tear resistance.
  • Black nitrile foam coating on palm and fingers provides oil impermeability, abrasion resistance, and good grip.
  • Tight knit provides comfort, support, dexterity, touch and limits dirt penetration.
  • Easily laundered for extended use.
  • Ventilated back optimizes breathability.
  • Elastic wrist.
  • Black color suitable for work in soiled environments.
  • TRUSTED_SUSTAINABILITY
    Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 Basic standards, EN 388-2003 Mechanical hazard, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number N°0075/797/162/05/07/0193, EC Attestation Number N°0075/797/162/05/07/0193
    Classification Dexterity: 5
    Coated Area Palm, Fingers
    Construction Seamless Knit
    Description Gauge 13
    Coating Material Nitrile Foam
    Color Black
    Cut Resistance Rating 1
    Material Nitrile, Polyamide
    Product Type Gloves
    Size 8
    For Use With (Application) Automotive, Aviation, Logistics, Maintenance and Transportation industries.
    Puncture Rating 1
    Quantity 10Pair/PK
    Tear Rating 2
    Length (Metric) 24.5 cm
    Show More Show Less
    missing translation for 'provideContentCorrection'

    missing translation for 'productTitle'