missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ Polythene Safety Barrier Tape

Brand:  Allsigns International™ PBT4

UNSPSC : 46161507

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12329707

  • £22.95 / Each


Estimated Shipment
07-07-2017
Add to basket

Specifications Description & Specifications

Specifications

Orange
25mm x 25m
Safety Barrier Tape
1Each
Polythene
Documentation

Technical Documents