missing translation for 'onlineSavingsMsg'
Learn More

Allsigns International™ Polythene Safety Barrier Tape

Brand:  Allsigns International™ PBT2

UNSPSC : 46161507

Code : 04

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12308837

  • £67.65 / Each


Estimated Shipment
09-06-2017
Add to basket

Specifications Description & Specifications

Specifications

Safety Barrier Tape
Polythene
1Each
75mm x 250m
Documentation

Technical Documents