Allsigns International™ Polythene Safety Barrier Tape


Overview
Brand: Allsigns International™

Manufacturer Part Number: PBT1

UNSPSC: 46161507

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12398827

Quantity Price
1 £ 68.92 / Each
Estimated Shipment
09-05-2017
Add to basket


Description and Specification

Specification

Quantity 1Each
Product Type Safety Barrier Tape
Material Polythene
Dimensions (L x W) 250m x 75mm
Documentation

Technical documents