missing translation for 'onlineSavingsMsg'
Learn More

Millipore™ Polytetrafluoroethylene Millex-FG

Brand:  Millipore™ SLFG75000

Code : 71

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10648261

  • £707.00 / Pack of 100


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Syringe
50mm
4.1 (60)bar(psi)
< 0.1mL
0.2μm
1/8 in. NPTM
No
121deg. C
Millex-FG
Polytetrafluoroethylene
Natural inlet/natural outlet
4000mL
19.6cm2
1/8 in. NPTM
4000mL
100/Pk.
Videos
Documentation