Learn More
Corning™ Polystyrene Stripette Pipets
Manufactured with optically clear polystyrene for accurate measurement. Corning Polystyrene Stripette™ Pipets are ideal for sterile tissue culture applications.
Brand:  Corning™ 7027
Code : 66
Additional Details : Weight : 7.00000kg
 View more versions of this product
Product Code. 10576043
  • £448.27 / Pack of 1000
Call for latest availability
+44 (0)1509 555 500
Add to basket
Specifications
Specifications
2mL
Stripette
1000Pack
1000/Pk.
Polystyrene
Videos