Learn More
Alfa Aesar™ Polystyrene standard, M.W. 65,000, Mw/Mn 1.06
Brand:  Alfa Aesar™ 041932.03
Code : NEW
Additional Details : CAS Number : 9003-53-6 Weight : 0.00100kg
Product Code. 15431747
  • £42.80 / 1g
Estimated Shipment date
from Supplier 05-03-2018
Add to basket
Specifications
Specifications
Polystyrene standard
9003-53-6
MFCD00084450
1g
M.W. 65,000, Mw/Mn 1.06
Powder
Odorless