Learn More
Thermo Scientific Alfa Aesar Polystyrene standard, M.W. 400,000, Mw/Mn 1.06
Brand: Thermo Scientific Alfa Aesar 041939.03
Polystyrene standard, M.W. 400,000, Mw/Mn 1.06
Code : NEW
Additional Details : CAS Number : 9003-53-6 Weight : 0.00100kg