missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene standard, M.W. 30,000, Mw/Mn 1.06, Thermo Scientific™

Catalog Number 11304917
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
9003-53-6
(C8H8)n
104.15
MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
*-CC(-*)C1=CC=CC=C1
This item is not returnable. View return policy
9003-53-6
(C8H8)n
104.15
MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
*-CC(-*)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Physical Form White powder
Quantity 1 g
IUPAC Name styrene
Odor Odorless
Chemical Name or Material Polystyrene standard, M.W. 30, 000, Mw/Mn 1.06
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'