missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polystyrene Petri Dishes
Clear. Fisherbrand™ Polystyrene Petri Dishes are perfect for microbial or cell culture applications.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 6.30000kg
Specifications
16.2 mm | |
Non-sterile | |
Petri Dish | |
3 |
90 mm | |
Polystyrene | |
600 Case |