missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One™ Polystyrene Petri Dishes

Manufactured from highly transparent polystyrene for its heat resistant properties to enable use with hot agar. Greiner BioOne™ Polystyrene Petri Dishes are available in a wide variety of formats and sizes to accommodate multiple research needs.

Brand:  Greiner Bio-One™ 8000078

Code : NEW

Additional Details : Weight : 26.00000kg

Product Code. 13219662

  • £227.00 / 25 L


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Heat resistant up to 60°C
  • Available in vented and non-vented
  • Choose from standard round design, square-profile dish, special models and contact dishes/OneWell Plate™ models
  • Easy stacking
  • Crystal polystyrene for optimal analysis by microscopy
Specifications

Specifications

35 mm
10 mm
Polystyrene
35 x 10mm
Yes
740 Pk.
Yes
Yes
Videos