missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Polystyrene, methacrylate terminated, M.W. 12,000, 33% w/w in cyclohexane

Copolymer used with acrylates and acrylamides; Impacts toughness, increases sheer strength

Brand:  Alfa Aesar™ 042402.AE

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 1993 Chem class : 1 Pack group : II

Quantity 100mL

 View more versions of this product

Product Code. 11344548

  • £21.00 / 100mL


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Polystyrene, methacrylate terminated
M.W. 12,000
MFCD00212587
UN1993

Specifications

Polystyrene, methacrylate terminated
0.864g/mL
MFCD00212587
1.476
100mL
M.W. 12,000
Liquid
UN1993
33% w/w in cyclohexane
Videos
SDS