missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polystyrene latex microsphere, 15.0 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
Description
Polystyrene latex microsphere is used as plastics, include protective packaging, containers, lids, bottles, trays, tumblers, and disposable cutlery. And also used in protein binding, size markers. The most popular use of latex beads is in the diagnostic use of adsorbed antibodies or antigens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 9003-53-6 |
Physical Form | Dispersion |
Molecular Formula | (C8H8)n |
MDL Number | MFCD00084450 |
Synonym | ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene |
Solubility Information | Soluble in benzene,chloroform,carbon tetrachloride,cyclohexane,dimethylformamide,methylene chloride,pyridine,tetrahydrofuran,toluene,xylene. Insoluble in acetone,butane,ethanol,ether,methanol,hexane,phenol,propanol,water. |
Quantity | 5 mL |
InChI Key | PPBRXRYQALVLMV-UHFFFAOYSA-N |
SMILES | *-CC(-*)C1=CC=CC=C1 |
IUPAC Name | styrene |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'