missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Polystop™ Dropping Bottles

Prevent chipping of the glass dropper upon reinsertion. Poulten Graf™ Polystop™ Dropping Bottles are available in two color varieties, a range of capacities and include an interchangeable plastic stopper.

Brand:  Poulten Graf™ L700/C/2

Code : 02

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12378208

  • £48.85 / Pack of 2


Estimated Shipment
29-03-2018
Add to basket

Description

Description

  • With interchangeable plastic stopper, vinyl bulb and graduated glass pipette
  • Available in clear or amber glass and a range of sizes
  • Narrow neck construction
Specifications

Specifications

500mL
0.2mL
Bottle, Dropping
Clear
10mL
Videos
Documentation