missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, Multi-Compendial, N.F., J.T.Baker™

Brand:  JT Baker 4116-12

Additional Details : Weight : 1.00000kg

Packaging Narrow Mouth Amber Glass Bottle, w/ Septum
Quantity 1L

 View more versions of this product

Product Code. 15568264

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

9005-64-5
522.676
443314
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
C26H50O10
HMFKFHLTUCJZJO-UHFFFAOYSA-N
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate

Specifications

Polysorbate 20
C26H50O10
HMFKFHLTUCJZJO-UHFFFAOYSA-N
2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
443314
148.00°C
1L = 1.11kg
9005-64-5
Narrow Mouth Amber Glass Bottle, w/ Septum
CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO
522.676
Multi-Compendial/NF
1L
Videos
Documents