Learn More
Fisherbrand™ Polypropylene Yellow Pipet Tip
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Yellow Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets.
£26.40 - £33.10
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10630574
|
Fisherbrand™
|
1 to 250 μL |
£33.10
Pack of 960 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
|||||
11537452
|
Fisherbrand™
|
1 to 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to the stable shape
- Autoclavable
- Guaranteed dust-free due to special ionization treatment
- Available pre-sterilized or non-sterile
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- Practice-oriented tip volumes
- Aeroject™ tips with special filters made of hydrophobic PE-HD as
- Aerosol barrier and for protection against contamination
- Suitable for all commonly used pipet types
Specifications
Yellow | |
Autoclavable | |
Polypropylene |