Fisherbrand™ Polypropylene Yellow Pipet Tip

Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Yellow Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets.

Overview
Brand: Fisherbrand™

UNSPSC: 41121607

Code: 41

Additional Details:
Additional Details: Weight: 0.54200kg



Product Code. 10630574

Quantity Price
1 £ 51.1 / Pack of 960
Estimated Shipment
12-05-2017
Add to basket


Description and Specification

Specification

Certifications/Compliance CE, VID Certified
Volume (Metric) 1 to 250μL
Material Polypropylene
For Use With (Equipment) Single or Multichannel Pippets
Product Type Pipette Tip
Length (Metric) 5.12cm
Autoclavable Yes
Color Yellow
Quantity 960Pack
For Use With (Application) Ideal for All Commercially Available Pippets
Sterilization Method Autoclavable