Learn More
Kartell™ Polypropylene Weighing Bottles
Catalog Number 10792561
Click to view available options
Capacity: 20mL
Reduce the tare weight correction with these light weight, autoclavable, reusable translucent bottles. Kartell™ Polypropylene Weighing Bottles allow for the precise and secure weighing of samples in the laboratory.
  • Airtight push on lids included
  • Highly resistant to acid attack
  • No internal shoulders to allow use of spatulas
  • Tops can be squeezed to assist pouring
  • Remove lid before autoclaving
  • Suitable for foodstuff