missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Polypropylene V-Bottom 384 Well Protein LoBind Microplate
For troublesome samples where protein concentrations are low, the Eppendorf LoBind plates ensure maximal sample recovery - protecting enzyme activity and facilitating downstream analysis.
Brand: Eppendorf™ 0030624300
Code : 96
Additional Details : Weight : 3.51000kg
Description
- Eppendorf LoBind material ensures excellent sample recovery for improved assay results
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
- Available in tube, microplate, and deepwell plate formats for easy-up scaling
- High-contrast Unique OptiTrack™ matrix: up to 30 % faster sample identification and fewer pipetting errors
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
- Raised well rims and a smooth surface ensure reliable sealing in plates
Specifications
Standard | |
White, Clear Bottom | |
Polypropylene | |
Conical Bottom (V-well) | |
80/Pk. |
16 x 24 | |
384-Well Protein LoBind Microplate | |
384 | |
80 |