Learn More
Kartell™ Polypropylene Universal Tip
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Pipette Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
Brand:  Kartell™ 0965700
Code : NEW
Additional Details : Weight : 1.58400kg
Product Code. 10651122
  • £90.15 / Pack of 960
Estimated Shipment: 20-05-2024
to see stock.
Description
Description
  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market
  • Disposable tips are also available racked