missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polypropylene Two and Three Tier Test Tube Racks

Manufactured from polypropylene for strength and durability. Kartell™ Polypropylene Two and Three Tier Test Tube Racks hold ten tubes and are lightweight and unbreakable.

Brand:  Kartell™ 370

Code : 02

Additional Details : Weight : 0.09588kg

 View more versions of this product

Product Code. 11378633

Quantity Price
1 £9.85 / Each
10 £9.01 / Each
In Stock 22

Add to basket


Description

Description

  • Unbreakable traditional pattern
  • Lightweight
  • Autoclavable to 120°C.
  • Indentations on base area hold test tubes in position
Specifications

Specifications

Yes
250 x 60mm
Polypropylene
Rack, 2 Tier
18mm
Test Tubes (10)
1 Pack
Videos
Documentation