Kartell™ Polypropylene Micropipet Tips

Designed with high clarity and excellent water-repellent properties to keep sample retention within the tip to a minimum. Kartell™ Polypropylene Micropipet Tips feature precise centering of the orifice at the end of the tip to guarantee accurate delivery of the sample.

Overview
Brand: Kartell™

Manufacturer Part Number: 940

X1000 PUNTAS GILSON 1-200U

UNSPSC: 41120000

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12041173

Quantity Price
1 £ 10.0 / Pack of 1000
Estimated Shipment
21-03-2017
Add to basket


Description and Specification

Specification

Material Virgin cadmium-free polypropylene
Volume (Metric) 5 to 200μL
Product Type Gilson™
Autoclavable Yes
Tip Style Bevelled
Packaging Bulk
Certifications/Compliance CE–IVD
For Use With (Equipment) Recommend use with Kartell™ automatic micropipets, will deliver excellent results with other micropipet brands
Color Neutral
Compatibility Gilson, Kartell multipet, Bevelled tips
Color Neutral
Autoclavable Yes
Compatible Pipettes Gilson, Kartell multipet, Bevelled tips
Packaging Quantity 1000Pack
Packaging Bulk
Quantity 1000/pk
Graduated Yes
For Use With (Application) Deliver excellent results

  • Cadmium-free polypropylene construction provides high clarity and water-repellent properties
  • Specially designed for Kartell automatic pipets, but deliver excellent results with most available micropipet brands 
  • Autoclavable to 121°C for 20 minutes
  • Precision delivery of sample, minimal sample retention with tip 
  • Packaging selection satisfies a variety of needs:
    • bulk bag
    • packed in racks
    • refills for reloading racks