Learn More
DURAN™ Laboratory Thread Adapters
Brand:  Duran 1168449
Code : A0
Additional Details : Weight : 0.02600kg
 View more versions of this product
Product Code. 10767214
  • £86.45 / Each
Estimated Shipment
10-07-2018
Add to basket
Specifications
Specifications
1Each
Thread Adapter
Polypropylene
Thread Adapter
Handling aggressive, gas generating media
GL32/GL45
Videos