Learn More
Lenz Laborglasinstrumente™ Plastic Straight Hose Connector
For connecting screw caps with bore. Lenz Laborglasinstrumente™ Plastic Straight Hose Connectors are made from polypropylene.
Brand:  Lenz Laborglasinstrumente™ 01350001
Code : 50
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 12303379
  • £60.80 / Pack of 10
Estimated Shipment
29-06-2018
Add to basket
Description
Description
  • Autoclavable
  • Available in two different thread sizes
  • Silicone sealing
Specifications
Specifications
Yes
Straight Hose Connection
10 Pack
Straight
Polypropylene
For screw caps with bore
GL18
Videos
Documentation