missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Tapered Straight Connectors

Manufactured from polypropylene for strength and durability. Kartell™ Tapered Straight Connectors are available to fit a wide range of tubing.

Brand:  Kartell™ 515

Code : 02

Additional Details : Weight : 0.05900kg

Product Code. 12369518

  • £38.89 / Pack of 10


In Stock 2
Add to basket

You may only order up to the available stock. For more details call customer services on 01509 555 500

Description

Description

  • Autoclavable
  • For straight connection of different diameter tubing
  • CE marked
  • IVD approved
Specifications

Specifications

14 to 16mm
10.5mm Bore
Yes
Natural
100 Pack
Polypropylene
Straight
CE IVD
Videos
Documentation