missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polypropylene Straight Connector for Tubing

Connect tubing of the same or similar sizes with a straight pattern connector. The Azlon™ polypropylene straight connector for tubing is autoclavable.

Brand:  Azlon CWH608

Code : 01

Additional Details : Weight : 0.07144kg

 View more versions of this product

Product Code. 10155274

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Straight pattern tubing connectors in white polypropylene
  • For connecting tubing of similar or different sizes
  • Tapered arms are rigid with steps to give firm grip
  • Autoclavable at 121°C with good chemical resistance
Specifications

Specifications

Connector
Polypropylene
Straight
121°C
Autoclavable
7 to 12 and 10 to 15mm I.D. tubing
White
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers