Learn More
Kartell™ Polypropylene Stirring Paddles
Constructed of polypropylene and steel for strength, durability and chemical resistance. Kartel™ Polypropylene Stirring Paddles provide a sturdy variety of paddles to fit any stirring need.
Brand: Kartell™ 0043200
Additional Details : Weight : 0.25000kg
Description
- Steel rod covered with polypropylene provides a strong connection to the stirrer
- Steel rods: 8 dia. x 350mmL fits standard overhead stirrer
- Polypropylene blades are premanently fixed to the rods for stability
- Fixed propeller for vortex stirring
- U shape for slower but more vigorous agitation
- Two sizes of swivel impellers enable the blades to be introduced through a small aperture then fully extend for stirring
Specifications
Collapsible paddle, fully extends during stirring | |
98.5 mm W | |
Stirring Paddle | |
98.5 mm |