Learn More
Grant Instruments™ Polypropylene Spheres
Pack of 300 polypropylene spheres
Brand: Grant Instruments™ PS20
Code : 57
Additional Details : Weight : 0.42300kg
Description
Pack of 300 polypropylene spheres
Useful alternative to a lid, minimises evaporation and heat loss whilst allowing easy access to vessels in the bath; particularly useful for tall vesselsSpecifications
Bath Accessories | |
Polypropylene |