missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Polypropylene sheet, 12.7mm (0.5 in.) thick

Brand:  Alfa Aesar™ 045215.HB

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 300x300mm

Product Code. 15482207

  • £31.10 / Each


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Specifications

Specifications

Polypropylene sheet
12.7mm (0.5 in.) thick
MFCD00084447
300x300mm
Videos
SDS