missing translation for 'onlineSavingsMsg'
Learn More

Brand™ Polypropylene Scoops

Brand:  Brand™ 139130

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12055292

  • £25.30 / Pack of 6


Estimated Shipment
26-06-2018
Add to basket

Specifications

Specifications

White
Polypropylene
Scoop
315mm
500mL
Videos