Learn More
Oakton™ Polypropylene Sample Containers
Prevent sample contamination with these polypropylene containers. Cole Parmer™ Polypropylene Sample Containers are resistant to acids, bases and most organic solvents.
Brand: Oakton™ 06100-12
Code : A0
Additional Details : Weight : 4.98600kg
Description
- Dual graduations in mL and oz.
- Manufactured of translucent polypropylene
- Lids fit tightly, sold separately
Specifications
500/Pk. | |
Sample Container | |
6.5 cm |
130 mL | |
Polypropylene |