missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polypropylene Reservoir Column Kit

Brand:  Macherey-Nagel™ 730382

Code : 50

Additional Details : Weight : 0.01000kg

Product Code. 10586531

  • £99.00 / Pack of 10
Estimated Shipment: 16-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

70 mL
1, 3, 6 mL Chromabond Polypropylene Columns
Adapter
Reservoir
Reservoir Column Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers