Learn More
BRAND™ Polypropylene Reservoir
Reservoirs allow for different volumes and different solutions to be used in the automated liquid handling process.
Brand: BRAND™ 701454
Additional Details : Weight : 0.66130kg
Description
- Pyramidal or flat bottom
- DNase-free
- RNase-free
Specifications
60 mL/Column | |
Non-sterile | |
Yes | |
Conical |
Polypropylene | |
60 mL | |
4 |