missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ Reducer Tubing Connectors, Plastic

Variously sized and shaped reducer tubing connectors. Ismatec™ Reducer Tubing Connectors, Plastic are available in different plastic compositions.

Brand:  Ismatec™ ISM573A

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10052852

Quantity Price
1 £55.84 / Pack of 10
10 £45.15 / Pack of 10
Estimated Shipment: 30-07-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

  • Reducer tubing connector, with varying inlet and outlet diameters.
  • Designed for Ismatec™ pump tubing.
  • Available in different sizes, shapes, and compositions.
Specifications

Specifications

Acetal Resin (POM, Polyoxymethylene)
For 4.0 and 6.0mm I.D. Tubing
4.0mm and 6.0mm I.D.
Reducer Tube Connector
10/Pk.
Straight
Videos
SDS
Documents