missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Polypropylene Pipette Tips

Use Socorex™ Polypropylene Pipette Tips when a whole volume range since there is a whole volume range from which to choose. They are available in various colors, are constructed of polypropylene and are autoclavable for cost effectiveness.

Brand:  Socorex™ 309.0010B

Code : 03

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10656161

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Supplied in loose in bags or racks with lids
  • Autoclavable at 121°C / 250°F
  • Compatible with other brands' tips
Specifications

Specifications

Pipette Tip
Natural
Polypropylene
1000 Pack
5μL
Regular
Rack
Videos