Learn More
BUCHI Polypropylene O-ring
Brand: BUCHI 046363
Additional Details : Weight : 0.25000kg
Specifications
O-Ring | |
1Each | |
37.69 x 3.53mm |
For use with Coupling Spare for Mini Spray | |
Polypropylene |