missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Micrewtube™ Polypropylene Microtubes with O-Ring Seal Screw Cap

Use these tubes for cryogenic work. Simport™ Micrewtube™ Polypropylene Microtubes with O-Ring Seal Screw Cap keep the integrity of small samples under even the most adverse conditions.

Brand:  Simport™ T332-4

UNSPSC : 41121702

Code : 46

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11576819

  • £166.00 / Pack of 1000


In Stock 1
Add to basket

Specifications

Description

  • Available as self standing or conical
  • O-ring secured in the top of the cap ensures a positive leakproof seal
  • Supplied with attachment loops and allow them to remain attached to the tube in order to prevent mix up and contamination
  • Can be centrifuged at up to 17, 000 x g
  • Will withstand temperatures from -196 to +121°C
Description & Specifications

Specifications

1.5mL
1000/Pk.
-196 to +121°C
Polypropylene
Self-standing
Documentation