Polypropylene Non-sterile Microtubes with O-Ring Seal


Overview
Brand: Simport

Manufacturer Part Number: T3325

UNSPSC: 41121702

Code: 46

Additional Details:
Additional Details: Weight: 1.37600kg



Product Code. 11752853

Quantity Price
1 £ 166.0 / Pack of 1000
In Stock 2
Add to basket


Description and Specification

Specification

Packaging 1000/Pk.
Temperature Range (Metric) -196 to +121°C

These tubes are available as self standing or conical and have an O-ring secured in the top of the cap ensuring a positive leakproof seal.
The caps are supplied with attachment loops and allow them to remain attached to the tube in order to prevent mix up and contamination. Can be centrifuged at up to 17,000xg. Will withstand temperatures from -196°C to 121°C.

Documentation